Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Acetyl-L-carnitine hydrochloride, ≥96.0% (HPLC); 96.0-104.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111014382 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4382 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4382 Supplier MilliporeSigma Supelco Supplier No. 9210750MG
Add to Cart
Add to Cart

Acetyl carnitine is one of the major acylcarnitines produced during the transport of long-chain fatty acids in the body.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5080-50-2
Physical Form Powder/Crystals
Linear Formula C9H17 NO4 · HCl
MDL Number MFCD00082230
Synonym (R)-3-Acetoxy-4-(trimethylammonio)butyrate hydrochloride; ALC; ALCAR; R-(-)-2-Acetyloxy-3-carboxy-N,N,N-trimethyl-1-propanaminium chloride
Type Analytical Standard
Quantity 50 mg
Formula Weight 239.70
Percent Purity ≥96.0% (HPLC); 96.0 to 104.0% (AT)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.