Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O-acetyl-L-serine | 5147-00-2 | MFCD00037771 | 98.0% | 147.13 g/mol | C5H9NO4 | 5 G
SDP

Catalog No. 5000247648
Encompass_Preferred

O-Acetylserine (O-acetyl-L-serine) is an alpha-amino acid intermediate involved in the biosynthesis of cysteine in bacteria and plants. Supplied for research and analytical use, it is used in metabolic pathway studies, enzymology, and as an analytical reference. The substance is a crystalline solid with defined storage recommendations for long-term stability.

  • Intermediate in cysteine biosynthesis.
  • Suitable as an analytical and research standard.
  • Molecular weight 147.13 g/mol.
  • Chemical formula C5H9NO4.
  • Typical purity 98.0%.
  • Available in laboratory solid pack sizes, including 5 g.
  • Store powder at -20°C for long-term stability.

Catalog No. 50-002-47648 Supplier Medchemexpress LLC Supplier No. HY1014095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-Acetylserine (O-acetyl-L-serine) is an alpha-amino acid intermediate involved in the biosynthesis of cysteine in bacteria and plants. Supplied for research and analytical use, it is used in metabolic pathway studies, enzymology, and as an analytical reference. The substance is a crystalline solid with defined storage recommendations for long-term stability.

  • Intermediate in cysteine biosynthesis.
  • Suitable as an analytical and research standard.
  • Molecular weight 147.13 g/mol.
  • Chemical formula C5H9NO4.
  • Typical purity 98.0%.
  • Available in laboratory solid pack sizes, including 5 g.
  • Store powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.