Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Acetyl-L-serine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A08341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08341G 1 g
A0834100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08341G Supplier TCI America Supplier No. A08341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66638-22-0
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00060169
InChI Key MGQOSZSPKMBSRW-WCCKRBBISA-N
Synonym o-acetyl-l-serine hydrochloride, s-3-acetoxy-2-aminopropanoic acid hydrochloride, o-acetyl-l-serine hcl, h-ser ac-oh.hcl, o-acetyl-l-serine-hydrogen chloride 1/1
PubChem CID 44630053
IUPAC Name (2S)-3-acetyloxy-2-aminopropanoic acid;hydrochloride
SMILES CC(=O)OCC(C(=O)O)N.Cl

Specifications

Melting Point 163°C
Color White
Quantity 1 g
Formula Weight 183.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Acetyl-L-serine Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.