Learn More
Medchemexpress LLC HY-101409 100mg Medchemexpress, O-Acetylserine CAS:5147-00-2 Purity:>98%
SDP
Catalog No. 501932115
Encompass_Preferred
Medchemexpress, HY-101409 100mg O-Acetylserine CAS:5147-00-2 O-Acetylserine (O-Acetyl-L-serine) is an intermediate in the biosynthesis of the amino acid cysteine in bacteria and plants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2115 Supplier Medchemexpress LLC Supplier No. HY101409100MG
Medchemexpress, HY-101409 100mg O-Acetylserine CAS:5147-00-2 O-Acetylserine (O-Acetyl-L-serine) is an intermediate in the biosynthesis of the amino acid cysteine in bacteria and plants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.