Learn More
Bachem o-Agatoxin IVa Trifluoroacetate
SDP
Supplier:  Bachem 4030173.0100
Encompass_Preferred
o-Agatoxin IVa Trifluoroacetate, H-Lys-Lys-Lys-Cys-Ile-Ala-Lys-Asp-Tyr-Gly-Arg-Cys-Lys-Trp-Gly-Gly-Thr-Pro-Cys-Cys-Arg-Gly-Arg-Gly-Cys-Ile-Cys-Ser-Ile-Met-Gly-Thr-Asn-Cys-Glu-Cys-Lys-Pro-Arg-Leu-Ile-Met-Glu-Gly-Leu-Gly-Leu-Ala-OH, 145017-83-0, C217H360N68O60S10, Mr 5202.33, 0.1mg. o-Agatoxin IVa is a peptide isolated from the venom of the American funnel web spider, Agelenopsis aperta. o-Aga IVa is a selective and potent blocker of the mammalian P-type voltage-dependent calcium channel.
Catalog No. 50-259-508