Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Anisidine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. A048925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A048925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A048925G Supplier TCI America Supplier No. A048925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 134-29-2
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00054334
InChI Key XCZCWGVXRBJCCD-UHFFFAOYSA-N
Synonym 2-Methoxyaniline Hydrochloride
PubChem CID 8638
IUPAC Name 2-methoxyaniline;hydrochloride
SMILES COC1=CC=CC=C1N.Cl

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 159.61
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material o-Anisidine Hydrochloride
EINECSNumber (3)-0682
RTECSNumber BZ6500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.