Learn More
Cayman Chemical O-ArachIdonyl EthnolamIn 10mg
SDP
Catalog No. 502894252
Encompass_Preferred
A constituent of human and rat brain in which the ethanolamine moiety is attached “backwards”, as an ester rather than as an amide as in AEA; has mixed agonist/antagonist activity at the CB1 receptor
Catalog No. 50-289-4252 Supplier Cayman Chemical Supplier No. 9105010 MG
A constituent of human and rat brain in which the ethanolamine moiety is attached “backwards”, as an ester rather than as an amide as in AEA; has mixed agonist/antagonist activity at the CB1 receptor