Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(Benzotriazol-1-yl)-N,N,N',N'-bis(pentamethylene)uronium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. B38165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38165G 5 g
B38161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38165G Supplier TCI America Supplier No. B38165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 206752-41-2
Molecular Formula C17H24F6N5OP
Molecular Weight (g/mol) 459.377
MDL Number MFCD00191769
InChI Key YNOBMGHLCWIWCL-UHFFFAOYSA-N
Synonym (Benzotriazol-1-yloxy)dipiperidinocarbenium Hexafluorophosphate, HBPipU
PubChem CID 10863473
IUPAC Name 1-[piperidin-1-ium-1-ylidene(piperidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
SMILES C1CCN(CC1)C(=[N+]2CCCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 199°C
Color White
Quantity 5 g
Formula Weight 459.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-(Benzotriazol-1-yl)-N,N,N′,N′-bis(pentamethylene)uronium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.