Learn More
Medchemexpress LLC O-(benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate | 94790-37-1 | MFCD00075445 | >=98.0% | 379.24 g·mol⁻¹ | C11H16F6N5OP | 1kg
SDP
Catalog No. 5000268523
Encompass_Preferred
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]
Catalog No. 50-002-68523 Supplier Medchemexpress LLC Supplier No. HYY09121KG
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]