Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O-Benzyl-D-Serine

Catalog No. 139101GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-910-1GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-910-1GF Supplier Sigma Aldrich Supplier No. 139101GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

O-Benzyl-D-Serine

Specifications

CAS 719-80-2
Density 1.066 g/mL at 25°C
Molecular Formula C14H15OP
Linear Formula (C6H5)2POC2H5
Quantity 1 g
MDL Number MFCD00009087
Synonym (R)-2-Amino-3-benzyloxypropionic acid
Molecular Weight (g/mol) 230.24 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.