Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzyl-L-serine Methyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B14501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14501G 1 g
B14505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14501G Supplier TCI America Supplier No. B14501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19525-87-2
Molecular Formula C11H16ClNO3
Molecular Weight (g/mol) 245.70
MDL Number MFCD00191312
InChI Key UDHSAWHYRXHQJP-UHFFFAOYNA-N
Synonym Methyl O-Benzyl-L-serinate Hydrochloride, H-Ser(Bzl)-OMe.HCl
PubChem CID 44630138
IUPAC Name methyl 2-amino-3-(benzyloxy)propanoate hydrochloride
SMILES Cl.COC(=O)C(N)COCC1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Formula Weight 245.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzyl-L-serine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.