Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzyl-L-serine 99.0+%, TCI America™
SDP

Catalog No. B08615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08615G 5 g
B08611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08615G Supplier TCI America Supplier No. B08615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4726-96-9
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00065937
InChI Key IDGQXGPQOGUGIX-VIFPVBQESA-N
Synonym o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine
PubChem CID 78457
IUPAC Name (2S)-2-amino-3-phenylmethoxypropanoic acid
SMILES C1=CC=C(C=C1)COCC(C(=O)O)N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzyl-L-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.