Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Butylhydroxylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B56865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56865G 5 g
B56861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56865G Supplier TCI America Supplier No. B56865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4490-82-8
Molecular Formula C4H12ClNO
Molecular Weight (g/mol) 125.596
MDL Number MFCD00487631
InChI Key HUYRNQWVAPCTQZ-UHFFFAOYSA-N
PubChem CID 12283956
IUPAC Name O-butylhydroxylamine;hydrochloride
SMILES CCCCON.Cl

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 5 g
Formula Weight 125.60
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material O-Butylhydroxylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.