Learn More
Medchemexpress LLC O=C(NC1=CC=C(C=C1)C#C)C2=C3N=C(C=C(N3N=C2)C)C | 1380716-06-2 | 98.0% | 290.32 g/mol | C17H14N4O | 5 MG
SDP
Catalog No. 5000227819
Encompass_Preferred
Small-molecule glucocerebrosidase (GCase) modulator supplied as a 5 mg sample. It acts as an activator and non-inhibitory chaperone (reported EC50 = 0.4 μM) and contains an alkyne functional group for click chemistry.
  • Modulates glucocerebrosidase activity and acts as a non-inhibitory chaperone.
  • Reported EC50 = 0.4 μM in GCase assays.
  • Contains an alkyne functional group for copper-catalyzed click chemistry.
  • High purity (98.0%) and characterized molecular weight (290.32 g/mol).
  • Provided in small-scale quantities for in vitro research applications.
Catalog No. 50-002-27819 Supplier Medchemexpress LLC Supplier No. HY1178615MG
Small-molecule glucocerebrosidase (GCase) modulator supplied as a 5 mg sample. It acts as an activator and non-inhibitory chaperone (reported EC50 = 0.4 μM) and contains an alkyne functional group for click chemistry.
  • Modulates glucocerebrosidase activity and acts as a non-inhibitory chaperone.
  • Reported EC50 = 0.4 μM in GCase assays.
  • Contains an alkyne functional group for copper-catalyzed click chemistry.
  • High purity (98.0%) and characterized molecular weight (290.32 g/mol).
  • Provided in small-scale quantities for in vitro research applications.