Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O=C(NC1=CC=CC(CN(C(C=CS2)=C2C(N3CCC)=O)C3=O)=C1)CC4=CC=CC=C4 | 2098776-03-3 | 98.4% | 433.52 g·mol⁻¹ | C24H23N3O3S | 5 MG
SDP

Catalog No. 5000221662
Encompass_Preferred

FEN1-IN-SC13 is a small-molecule inhibitor of flap endonuclease 1 (FEN1) that interferes with DNA replication and repair and has reported antitumor activity in vitro and in cells. It is supplied for research use as a characterized solid or as a 10 mM solution in DMSO, with documented purity and storage recommendations.

  • Potent FEN1 inhibition with reported antitumor activity.
  • Purity 98.42%.
  • Chemical formula C24H23N3O3S; molecular weight 433.52 g·mol⁻¹.
  • Available as solid (5-500 mg) and 10 mM 1 mL solution in DMSO.
  • Store powder at -20°C (long term) or 4°C (short term); store solutions at -80°C or -20°C as recommended.
  • Intended for research use only.

Catalog No. 50-002-21662 Supplier Medchemexpress LLC Supplier No. HY1457585MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FEN1-IN-SC13 is a small-molecule inhibitor of flap endonuclease 1 (FEN1) that interferes with DNA replication and repair and has reported antitumor activity in vitro and in cells. It is supplied for research use as a characterized solid or as a 10 mM solution in DMSO, with documented purity and storage recommendations.

  • Potent FEN1 inhibition with reported antitumor activity.
  • Purity 98.42%.
  • Chemical formula C24H23N3O3S; molecular weight 433.52 g·mol⁻¹.
  • Available as solid (5-500 mg) and 10 mM 1 mL solution in DMSO.
  • Store powder at -20°C (long term) or 4°C (short term); store solutions at -80°C or -20°C as recommended.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.