Learn More
VWR INTERNATIONAL INC O.C.T COMPOUND, SAKURA TISSUE-
Catalog No. NC1862249
Encompass
O.C.T Compound, sakura Tissue-Tek, , Embedding Medium for Frozen Tissue Specimens to ensure Optimal Cutting
This item is not returnable. View return policy
Catalog No. NC1862249 Supplier VWR INTERNATIONAL INC Supplier No. 25608930
O.C.T Compound, sakura Tissue-Tek, , Embedding Medium for Frozen Tissue Specimens to ensure Optimal Cutting