Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O-(Carboxymethyl)hydroxylamine hemihydrochloride

Catalog No. C1340825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1340825G 25 g
C1340810G 10 g
C134081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C1340825G Supplier Sigma Aldrich Supplier No. C1340825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

O-(Carboxymethyl)hydroxylamine hemihydrochloride

Specifications

CAS 2921-14-4
Melting Point 156°C (lit.)
Molecular Formula C2H5NO3 -+ 0.5 HCl
Linear Formula NH2OCH2COOH -+ 0.5 HCl
MDL Number MFCD00012955
Quantity 25 g
Synonym (Aminooxy)acetic acid hemihydrochloride; (Carboxymethoxy)amine hemihydrochloride; Hydroxylamine-O-acetic acid hemihydrochloride
Molecular Weight (g/mol) 109.3
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.