Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ O'Cedar™ Outdoor Power Corner Broom

Catalog No. 19200799 Shop All Vileda Professional Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-200-799 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-799 Supplier Vileda Professional™ Supplier No. 159833

O'Cedar™ Outdoor Power Corner Broom

  • Eco-friendly bristles are made from 80% recycled plastic bottles and a block made from 50% recycled plastic
  • Features foam comfort grip for ease of use
  • Angled bristles are strong enough to sweep heavy debris

Specifications

Product Type Broom
Color Red, Black
Sterility Non-sterile
Autoclavable Not Autoclavable
Quantity 4/Cs.
Description Outdoor Angle Broom
For Use With (Application) Garages, Patios and Sidewalks
Material Aluminum, Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.