Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ O-Cedar ™ Scrunge Heavy Duty Scrubber Sponge

Catalog No. 19200745 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Blue, Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-200-745 Blue, Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-745 Supplier Vileda Professional™ Supplier No. 148377
  • $15.50
    $10.65 / Pack of 2

    Save $4.85

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $124.00
    $85.65 / Case of 8 PK

    Save $38.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Scour away tough dirt, grease and grime with this heavy-duty cleaning sponge. The rippled scrubber surface rinses clean versus traditional scrubbers that trap and collect dirt.

  • Rippled scrubber surface breaks up and lifts away dirt then rinses clean to prevent odors
  • Built with durable material that lasts 20% longer than regular scrubbing sponges
  • Extra thick and thirsty sponge quickly wipes surfaces clean

Specifications

Product Type Heavy Duty Sponge
Sterility Non-sterile
Color Blue, Black
Dimensions (L x W x H) 4.5 x 3 x 1.5 in.
For Use With (Application) Counter or table tops
Height (Metric) 3.8 cm
Length (Metric) 11.5 cm
Material Cellulose
Quantity 2/Pk., 8Pk./Cs.
Width (Metric) 7.6 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.