Learn More
Medchemexpress LLC O-(chloroacetyl-carbamoyl) fumagillol | 129298-91-5 | MFCD00870281 | >98.0% | 401.9 | C19H28ClNO6 | 1 MG
SDP
Catalog No. 5000249011
Encompass_Preferred
TNP-470 is a synthetic analog of fumagillin that irreversibly inhibits methionine aminopeptidase-2 (MetAP2), blocking endothelial cell proliferation and angiogenesis. It is supplied as a crystalline solid for research use, typically at >98% purity and stored at -20 °C. Common applications include studies of angiogenesis, MetAP2-related signaling, and preclinical evaluation of anti-angiogenic strategies.
  • Irreversible MetAP2 inhibitor with anti-angiogenic activity.
  • High purity suitable for biochemical and cell-based assays.
  • Crystalline solid form for accurate dosing and stability.
  • Storage at -20 °C maintains compound integrity.
  • Used in vascular biology and cancer research models.
Catalog No. 50-002-49011 Supplier Medchemexpress LLC Supplier No. HY1019321MG
TNP-470 is a synthetic analog of fumagillin that irreversibly inhibits methionine aminopeptidase-2 (MetAP2), blocking endothelial cell proliferation and angiogenesis. It is supplied as a crystalline solid for research use, typically at >98% purity and stored at -20 °C. Common applications include studies of angiogenesis, MetAP2-related signaling, and preclinical evaluation of anti-angiogenic strategies.
  • Irreversible MetAP2 inhibitor with anti-angiogenic activity.
  • High purity suitable for biochemical and cell-based assays.
  • Crystalline solid form for accurate dosing and stability.
  • Storage at -20 °C maintains compound integrity.
  • Used in vascular biology and cancer research models.