Learn More
Bachem o-Conotoxin MVIIA
SDP
Supplier:  Bachem 4025279.1000
Encompass_Preferred
o-Conotoxin MVIIA , H-Cys-Lys-Gly-Lys-Gly-Ala-Lys-Cys-Ser-Arg-Leu-Met-Tyr-Asp-Cys-Cys-Thr-Gly-Ser-Cys-Arg-Ser-Gly-Lys-Cys-NH2, 107452-89-1, C102H172N36O32S7, Mr 2639.17, 1mg. o-Conotoxin MVIIA, originally isolated from the venom of the fish-hunting cone snail Conus magus, is a blocker of voltage-sensitive Ca2+ channels in neurons. The peptide has been used to identify different Ca2+ channel subtypes in amphibian brain.
Catalog No. 50-260-182