Learn More
Medchemexpress LLC o-cresol-d7 | 202325-50-6 | MFCD03428136 | 95.0% | 115.18 | C7HD7O | 25 MG
SDP
Catalog No. 5000284377
Encompass_Preferred
o-Cresol-d7 is the deuterium-labeled form of o-cresol supplied as a stable isotope standard for use in mass spectrometry and metabolite studies. It serves as an internal standard and tracer for quantitative analyses of cresol metabolites and related biochemical pathways.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for quantitative analysis of cresol metabolites.
  • Available in small milligram pack sizes for trace-level work.
  • Includes product datasheet and handling instructions for laboratory use.
Catalog No. 50-002-84377 Supplier Medchemexpress LLC Supplier No. HYW65371825MG
o-Cresol-d7 is the deuterium-labeled form of o-cresol supplied as a stable isotope standard for use in mass spectrometry and metabolite studies. It serves as an internal standard and tracer for quantitative analyses of cresol metabolites and related biochemical pathways.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for quantitative analysis of cresol metabolites.
  • Available in small milligram pack sizes for trace-level work.
  • Includes product datasheet and handling instructions for laboratory use.