Learn More
Medchemexpress LLC o-cresol-d7 | 202325-50-6 | MFCD03428136 | 98.0% | 115.18 | C7HD7O | 10 MG
SDP
Catalog No. 5000284715
Encompass_Preferred
o-Cresol-d7 is the deuterium-labeled analog of o-cresol used as an isotope-labeled reference standard for analytical and research applications. It is commonly used as an internal standard or tracer in mass spectrometry, GC-MS, and LC-MS, and for quantitative metabolite and biomarker studies.
  • Deuterated internal standard (d7) for MS, GC-MS, and LC-MS.
  • High isotopic enrichment, typically 98.0% atom D.
  • Molecular formula C7HD7O and molecular weight 115.18.
  • Provided in small, pre-weighed quantities suitable for calibration and method development.
  • Suitable for metabolomics, toxicology, and quantitative analytical workflows.
Catalog No. 50-002-84715 Supplier Medchemexpress LLC Supplier No. HYW65371810MG
o-Cresol-d7 is the deuterium-labeled analog of o-cresol used as an isotope-labeled reference standard for analytical and research applications. It is commonly used as an internal standard or tracer in mass spectrometry, GC-MS, and LC-MS, and for quantitative metabolite and biomarker studies.
  • Deuterated internal standard (d7) for MS, GC-MS, and LC-MS.
  • High isotopic enrichment, typically 98.0% atom D.
  • Molecular formula C7HD7O and molecular weight 115.18.
  • Provided in small, pre-weighed quantities suitable for calibration and method development.
  • Suitable for metabolomics, toxicology, and quantitative analytical workflows.