Learn More
Medchemexpress LLC O-cresol-d7 | 202325-50-6 | MFCD03428136 | >98.0% | 115.18 | C7HD7O | 5 MG
SDP
Catalog No. 5000284421
Encompass_Preferred
o-Cresol-d7 is the deuterium-labeled isotopologue of o-cresol used as an internal standard and analytical reference for mass spectrometry and metabolic biomarker research. The material is supplied in a 5 mg quantity and is intended for analytical, calibration, and method-development applications.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for quantitative analysis, method development, and biomarker studies.
  • Small, pre-measured 5 mg quantity for analytical workflows.
  • High isotopic enrichment (98 atom % D) and chemical purity ≥98% reported by standards suppliers.
  • CAS number 202325-50-6; molecular formula C7HD7O; molecular weight 115.18.
Catalog No. 50-002-84421 Supplier Medchemexpress LLC Supplier No. HYW6537185MG
o-Cresol-d7 is the deuterium-labeled isotopologue of o-cresol used as an internal standard and analytical reference for mass spectrometry and metabolic biomarker research. The material is supplied in a 5 mg quantity and is intended for analytical, calibration, and method-development applications.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for quantitative analysis, method development, and biomarker studies.
  • Small, pre-measured 5 mg quantity for analytical workflows.
  • High isotopic enrichment (98 atom % D) and chemical purity ≥98% reported by standards suppliers.
  • CAS number 202325-50-6; molecular formula C7HD7O; molecular weight 115.18.