Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Cresolphthalein Complexone, Sigma-Aldrich

Catalog No. 01258911 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-911 100 g
01-258-874 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-911 Supplier MilliporeSigma Supelco Supplier No. P5631100G
Add to Cart
Add to Cart

o-Cresolphthalein Complexone has been used in the estimation of calcium in serum samples based on the reaction of Ca2+ with o-cresolphthalein complexone in an alkaline solution.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2411-89-4
Molecular Weight (g/mol) 636.6 g/mol

Specifications

Color White to Off-white
Quantity 100 g
Physical Form Powder
Chemical Name or Material o-Cresolphthalein Complexone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.