Learn More
Cayman Chemical O-Demethyl Apremilast, Purity: ≥95%, CAS: 1384441-38-6, 10 mg
SDP
Catalog No. 502948454
Encompass_Preferred
An active metabolite of apremilast; inhibits the activity of PDE4 isolated from U937 cells and LPS-induced TNF-α production in isolated PBMCs (IC50s = 8.3 and 5.6 µM, respectively); an oxidative degradation product of apremilast
Catalog No. 50-294-8454 Supplier Cayman Chemical Supplier No. 3421610 MG
An active metabolite of apremilast; inhibits the activity of PDE4 isolated from U937 cells and LPS-induced TNF-α production in isolated PBMCs (IC50s = 8.3 and 5.6 µM, respectively); an oxidative degradation product of apremilast