Learn More
Selleck Chemical LLC O-Demethyl-Gefitinib-E0833-5MG
SDP
Catalog No. 503137872
Encompass_Preferred
O-Desmethyl gefitinib a major active metabolite of gefitinib is a potent EGFR (ErbB1/HER1) inhibitor inhibits subcellular EGFR tyrosine kinase through a mechanism similar to that of gefitinib with IC50 values of 36 and 22 nM respectively
Catalog No. 50-313-7872 Supplier Selleck Chemical LLC Supplier No. E08335MG
O-Desmethyl gefitinib a major active metabolite of gefitinib is a potent EGFR (ErbB1/HER1) inhibitor inhibits subcellular EGFR tyrosine kinase through a mechanism similar to that of gefitinib with IC50 values of 36 and 22 nM respectively