Learn More
Medchemexpress LLC O-demethyl lenvatinib hydrochloride | 00-00-0 | 97.1% | C20H18Cl2N4O4 | 10MG
SDP
Catalog No. 5000259485
Encompass_Preferred
O-demethyl lenvatinib hydrochloride is the hydrochloride salt of the O-demethyl metabolite of lenvatinib, supplied for research use as a solid or as a 10 mM solution in DMSO. It is intended as a reference standard and tool compound for biochemical, cellular, and metabolism studies. Typical listed purity is 97.1% and molecular weight (salt) is 449.29 g·mol-1.
  • Hydrochloride salt form for improved solubility in polar solvents.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity suitable for analytical and reference standard use.
  • Applicable to metabolism, pharmacokinetic, and biochemical studies.
  • Supplied in small pack sizes suitable for research-scale experiments.
Catalog No. 50-002-59485 Supplier Medchemexpress LLC Supplier No. HY133980A10MG
O-demethyl lenvatinib hydrochloride is the hydrochloride salt of the O-demethyl metabolite of lenvatinib, supplied for research use as a solid or as a 10 mM solution in DMSO. It is intended as a reference standard and tool compound for biochemical, cellular, and metabolism studies. Typical listed purity is 97.1% and molecular weight (salt) is 449.29 g·mol-1.
  • Hydrochloride salt form for improved solubility in polar solvents.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity suitable for analytical and reference standard use.
  • Applicable to metabolism, pharmacokinetic, and biochemical studies.
  • Supplied in small pack sizes suitable for research-scale experiments.