Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O-demethyl lenvatinib hydrochloride | 97.1% | 449.29 g/mol | C20H18Cl2N4O4 | 5 MG
SDP

Catalog No. 5000262710
Encompass_Preferred

O-demethyl lenvatinib hydrochloride is the O-demethylated metabolite of the kinase inhibitor lenvatinib, provided as a hydrochloride salt for research and analytical applications. It is intended as an analytical standard for in vitro studies, method development, and metabolite identification.

  • Metabolite of lenvatinib for metabolite profiling and analytical reference.
  • Hydrochloride salt form for improved stability and handling.
  • High purity (97.1%) suitable for analytical workflows.
  • Small 5 mg pack size ideal for method development and reference use.
  • Molecular weight 449.29 g/mol and formula C20H18Cl2N4O4 for accurate calculations.

Catalog No. 50-002-62710 Supplier Medchemexpress LLC Supplier No. HY133980A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-demethyl lenvatinib hydrochloride is the O-demethylated metabolite of the kinase inhibitor lenvatinib, provided as a hydrochloride salt for research and analytical applications. It is intended as an analytical standard for in vitro studies, method development, and metabolite identification.

  • Metabolite of lenvatinib for metabolite profiling and analytical reference.
  • Hydrochloride salt form for improved stability and handling.
  • High purity (97.1%) suitable for analytical workflows.
  • Small 5 mg pack size ideal for method development and reference use.
  • Molecular weight 449.29 g/mol and formula C20H18Cl2N4O4 for accurate calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.