Learn More
Cayman Chemical O-desethyl DapaglIflozIn 10mg
SDP
Catalog No. 502911627
Encompass_Preferred
An SGLT2 inhibitor (IC50 = 9.4 nM) and active metabolite of dapagliflozin; selective for SGLT2 over SGLT1 (IC50 = 970 nM)
Catalog No. 50-291-1627 Supplier Cayman Chemical Supplier No. 3602210 MG
An SGLT2 inhibitor (IC50 = 9.4 nM) and active metabolite of dapagliflozin; selective for SGLT2 over SGLT1 (IC50 = 970 nM)