Learn More
Medchemexpress LLC O-desmethyl galanthamine | 60755-80-8 | MFCD07369289 | 99.7% | C16H19NO3 | 10MG
SDP
Catalog No. 5000260891
Encompass_Preferred
O-desmethyl galanthamine (sanguinine) is a galanthamine-type alkaloid used as a research reagent and reference standard. It acts as a reversible acetylcholinesterase (AChE) inhibitor and is used in biochemical assays to evaluate AChE activity and inhibitor potency; reported IC50 = 1.83 μM. Supplied as a powder for analytical and research use.
  • Reversible acetylcholinesterase inhibitor with reported IC50 1.83 μM.
  • High purity (99.7%) suitable for analytical applications.
  • Suitable as a reference standard in biochemical and pharmacological assays.
  • Powder form compatible with common laboratory solvents.
  • Molecular formula C16H19NO3; molecular weight 273.33 g·mol⁻¹.
  • Available in small quantities for research use.
Catalog No. 50-002-60891 Supplier Medchemexpress LLC Supplier No. HY13141310MG
O-desmethyl galanthamine (sanguinine) is a galanthamine-type alkaloid used as a research reagent and reference standard. It acts as a reversible acetylcholinesterase (AChE) inhibitor and is used in biochemical assays to evaluate AChE activity and inhibitor potency; reported IC50 = 1.83 μM. Supplied as a powder for analytical and research use.
  • Reversible acetylcholinesterase inhibitor with reported IC50 1.83 μM.
  • High purity (99.7%) suitable for analytical applications.
  • Suitable as a reference standard in biochemical and pharmacological assays.
  • Powder form compatible with common laboratory solvents.
  • Molecular formula C16H19NO3; molecular weight 273.33 g·mol⁻¹.
  • Available in small quantities for research use.