Learn More
Medchemexpress LLC O-desmethyl galanthamine (sanguinine) | 60755-80-8 | MFCD07369289 | 99.7% | 273.33 g/mol | C16H19NO3 | 5 MG
SDP
Catalog No. 5000262805
Encompass_Preferred
O-Desmethyl Galanthamine (Sanguinine) is a galanthamine-type alkaloid and an acetylcholinesterase (AChE) inhibitor used in neuropharmacology and biochemical research. It is provided as a high-purity powder and in DMSO solution formats for in vitro assays and mechanistic studies.
  • High purity suitable for research-grade assays.
  • Validated acetylcholinesterase inhibition; reported IC50 1.83 μM.
  • Available as powder and DMSO solutions for flexible preparation.
  • Stable when stored under recommended conditions for extended shelf life.
  • Supplied in small quantities appropriate for screening and mechanistic work.
Catalog No. 50-002-62805 Supplier Medchemexpress LLC Supplier No. HY1314135MG
O-Desmethyl Galanthamine (Sanguinine) is a galanthamine-type alkaloid and an acetylcholinesterase (AChE) inhibitor used in neuropharmacology and biochemical research. It is provided as a high-purity powder and in DMSO solution formats for in vitro assays and mechanistic studies.
  • High purity suitable for research-grade assays.
  • Validated acetylcholinesterase inhibition; reported IC50 1.83 μM.
  • Available as powder and DMSO solutions for flexible preparation.
  • Stable when stored under recommended conditions for extended shelf life.
  • Supplied in small quantities appropriate for screening and mechanistic work.