Learn More
Cayman Chemical O-desmethyl Galantamine, Purity: ≥90%, CAS: 60755-80-8, 10 mg
SDP
Catalog No. 502910634
Encompass_Preferred
An active metabolite of galantamine; inhibits erythrocyte and brain AChE and plasma BChE (IC50s = 0.12, 0.5, and 24 µM, respectively); prevents decreases in the viability of SH-SY5Y cells induced by hydrogen peroxide, amyloid-β (25-35), or cobalt chloride at 6.25-100 µM
Catalog No. 50-291-0634 Supplier Cayman Chemical Supplier No. 3508010 MG
An active metabolite of galantamine; inhibits erythrocyte and brain AChE and plasma BChE (IC50s = 0.12, 0.5, and 24 µM, respectively); prevents decreases in the viability of SH-SY5Y cells induced by hydrogen peroxide, amyloid-β (25-35), or cobalt chloride at 6.25-100 µM