Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O-desmethyl midostaurin | 740816-86-8 | 97.1% | 556.61 g/mol | C34H28N4O4 | 5 MG
SDP

Catalog No. 5000262339
Encompass_Preferred

O-desmethyl midostaurin is the primary active metabolite of the kinase inhibitor midostaurin. Supplied as a solid research reagent, it is intended for in vitro and preclinical research use only and is not for human or clinical use. The compound is characterized by a molecular formula of C34H28N4O4 and a molecular weight of 556.61 g/mol.

  • Active metabolite used in metabolism and pharmacology studies.
  • Molecular formula C34H28N4O4 and molecular weight 556.61 g/mol.
  • Purity 97.1%.
  • Off-white to light yellow solid powder.
  • Store powder at -20°C for long-term stability; short-term storage at 4°C.
  • Stable in solvent at -80°C for up to 6 months.
  • For research use only; not intended for human or clinical use.

Catalog No. 50-002-62339 Supplier Medchemexpress LLC Supplier No. HY1294915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-desmethyl midostaurin is the primary active metabolite of the kinase inhibitor midostaurin. Supplied as a solid research reagent, it is intended for in vitro and preclinical research use only and is not for human or clinical use. The compound is characterized by a molecular formula of C34H28N4O4 and a molecular weight of 556.61 g/mol.

  • Active metabolite used in metabolism and pharmacology studies.
  • Molecular formula C34H28N4O4 and molecular weight 556.61 g/mol.
  • Purity 97.1%.
  • Off-white to light yellow solid powder.
  • Store powder at -20°C for long-term stability; short-term storage at 4°C.
  • Stable in solvent at -80°C for up to 6 months.
  • For research use only; not intended for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.