Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Desmethylnaproxen, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018421 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8421 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8421 Supplier MilliporeSigma Supelco Supplier No. 9511425MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 52079-10-4
Physical Form Neat
Linear Formula C13H12O3
MDL Number MFCD00869765
Synonym (S)-6-Hydroxy-alpha-methyl-2-naphthaleneacetic acid; d-2-(6-Hydroxy-2-naphthyl)proprionic acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 216.23
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.