Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Desmethylvenlafaxine, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015927 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5927 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5927 Supplier MilliporeSigma Supelco Supplier No. 9386750MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-Desmethylvenlafaxine is a major active metabolite of the drug venlafaxine.

TRUSTED_SUSTAINABILITY

Specifications

CAS 93413-62-8
Physical Form Neat
Linear Formula C16H25 NO2
MDL Number MFCD00871934
Synonym 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol; Desvenlafaxine
Type Analytical Standard
Quantity 50 mg
Formula Weight 263.38
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.