Learn More
Sigma Aldrich Fine Chemicals Biosciences o-Dianisidine dihydrochloride tablet, 10 mg substrate per tablet | 20325-40-0 | MFCD00012488 | 50TAB
Catalog No. 501805528
Encompass
o-Dianisidine dihydrochloride tablet, 10 mg substrate per tablet | Mol Wt: 317.21 | 20325-40-0 | MFCD00012488 | 50TAB
Catalog No. 50-180-5528 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D915450TAB
o-Dianisidine dihydrochloride tablet, 10 mg substrate per tablet | Mol Wt: 317.21 | 20325-40-0 | MFCD00012488 | 50TAB