Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Dianisidine, free base, 99.9%, MP Biomedicals™

Catalog No. ICN10153625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10153625 25 g
ICN10153601 1 g
ICN10153605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10153625 Supplier MP Biomedicals, Inc Supplier No. 0210153625
Add to Cart
Add to Cart

Chemical Identifiers

CAS 119-90-4
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.294
InChI Key JRBJSXQPQWSCCF-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine, o-dianisidine, dianisidine, cellitazol b, neutrosel navy bn, diacel navy dc, disperse black 6, blue base nb, blue bn base, blue base irga b
PubChem CID 8411
ChEBI CHEBI:82321
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N

Specifications

Color Brown
Assay Percent Range 99.9%
Quantity 25 g
Formula Weight 244.3
Physical Form Powder
Chemical Name or Material o-Dianisidine, free base
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.