Learn More
Medchemexpress LLC O-Ethylhydroxylamine-d5 hydrochloride | 118087-07-3 | 99.7% | 102.57 | 5 MG
SDP
Catalog No. 5000452309
Encompass_Preferred
O-Ethylhydroxylamine-d5 (hydrochloride) is a deuterium-labeled form of O-Ethylhydroxylamine hydrochloride. It serves as a valuable tool for research, functioning as a tracer and an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, like deuterium, into drug molecules is a growing area of study, primarily utilized for quantitative analysis during the drug development process. Deuteration specifically holds promise for modifying the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
  • Used as a tracer for quantitative analysis.
  • Functions as an internal standard in analytical techniques.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Contains stable heavy isotopes for research in drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52309 Supplier Medchemexpress LLC Supplier No. HYW010619S5MG
O-Ethylhydroxylamine-d5 (hydrochloride) is a deuterium-labeled form of O-Ethylhydroxylamine hydrochloride. It serves as a valuable tool for research, functioning as a tracer and an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, like deuterium, into drug molecules is a growing area of study, primarily utilized for quantitative analysis during the drug development process. Deuteration specifically holds promise for modifying the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
  • Used as a tracer for quantitative analysis.
  • Functions as an internal standard in analytical techniques.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Contains stable heavy isotopes for research in drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.