Learn More
Enzo Life Sciences O-GlcNAc monoclonal antibody (CTD110.6) (purified) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS112A0100
Encompass_Preferred
Clone: CTD110.6. Isotype: IgM. Immunogen: Synthetic peptide containing serine-O-GlcNAc. Species reactivity: Species independent. Specificity: Recognizes Ser-O-GlcNAc and Thr-O-GlcNAc in eukaryotes. Interaction between the antibody and the Ser- or Thr-O-GlcNAc epitope can be blocked by competition with free GlcNAc. Applications: IF, IHC (PS). Purity: Purified by affinity chromatography. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-2669