Learn More
BIOHIPPO O-GlcNAc rAb W-R17
SDP
Catalog No. 5000975464
Encompass
U-Blot Pan-O-GlcNAc Rabbit mAb-BSA and Azide free is a Rabbit-derived recombinant monoclonal antibody (clone W-R17) that specifically recognizes O-GlcNAc a o-glcnacylation modification at Ser(S)/Thr(T) It shows reactivity with Human Mouse Rat Validated applications include WB Recommended dilutions WB 11000 Purified by protein a O-GlcNAc (short for O-linked -N-acetylglucosamine) is a reversible enzymatic post-translational modification found on serine and threonine residues of nucleocytoplasmic proteins The modification is characterized by a -glycosidic bond between the hydroxyl group of serine or threonine side chains and N-acetylglucosamine (GlcNAc) It is widely involved in the regulation of diverse cellular processes including transcription cell cycle signal transduction and stress response Dysregulation of O-GlcNAcylation has been closely linked to cancer neurodegeneration diabetes and inflammatory diseases Supplied as 100 uL
Catalog No. 50-009-75464 Supplier BIOHIPPO Supplier No. BHA21821407
U-Blot Pan-O-GlcNAc Rabbit mAb-BSA and Azide free is a Rabbit-derived recombinant monoclonal antibody (clone W-R17) that specifically recognizes O-GlcNAc a o-glcnacylation modification at Ser(S)/Thr(T) It shows reactivity with Human Mouse Rat Validated applications include WB Recommended dilutions WB 11000 Purified by protein a O-GlcNAc (short for O-linked -N-acetylglucosamine) is a reversible enzymatic post-translational modification found on serine and threonine residues of nucleocytoplasmic proteins The modification is characterized by a -glycosidic bond between the hydroxyl group of serine or threonine side chains and N-acetylglucosamine (GlcNAc) It is widely involved in the regulation of diverse cellular processes including transcription cell cycle signal transduction and stress response Dysregulation of O-GlcNAcylation has been closely linked to cancer neurodegeneration diabetes and inflammatory diseases Supplied as 100 uL