Learn More
New England Biolabs, Inc. O-Glycosidase – 10000000 units
SDP
Catalog No. 508112033
Encompass
O-Glycosidase, also known as Endo--N-Acetylgalactosaminidase, catalyzes the removal of Core 1 and Core 3 O-linked disaccharides from glycoproteins.
Catalog No. 50-811-2033 Supplier New England Biolabs, Inc. Supplier No. P0733L
O-Glycosidase, also known as Endo--N-Acetylgalactosaminidase, catalyzes the removal of Core 1 and Core 3 O-linked disaccharides from glycoproteins.