Learn More
New England Biolabs, Inc. O-Glycosidase – 2000000 units
SDP
Catalog No. 508112034
Encompass
O-Glycosidase, also known as Endo--N-Acetylgalactosaminidase, catalyzes the removal of Core 1 and Core 3 O-linked disaccharides from glycoproteins.
Catalog No. 50-811-2034 Supplier New England Biolabs, Inc. Supplier No. P0733S
Add to Cart
Add to Cart
O-Glycosidase, also known as Endo--N-Acetylgalactosaminidase, catalyzes the removal of Core 1 and Core 3 O-linked disaccharides from glycoproteins.