Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O-Linked GlcNAc transferase substrate | 954381-93-2 | 98.25% | 1783.08 | 10 MG
SDP

Catalog No. 5000398243
Encompass_Preferred

O-Linked GlcNAc transferase substrate is a biologically active peptide that functions as a substrate for O-linked GlcNAc transferase (OGT), a eukaryotic glycosyltransferase that utilizes UDP-GlcNAc as a glycosyl donor.

  • Biological active peptide
  • Substrate of O-linked GlcNAc transferase (OGT)
  • White to off-white solid appearance
  • High purity of 98.25%
  • Soluble in DMSO and H2O at 100 mg/mL
  • Sequence: Lys-Lys-Lys-Tyr-Pro-Gly-Gly-Ser-Thr-Pro-Val-Ser-Ser-Ala-Asn-Met-Met

Catalog No. 50-003-98243 Supplier Medchemexpress LLC Supplier No. HYP537510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-Linked GlcNAc transferase substrate is a biologically active peptide that functions as a substrate for O-linked GlcNAc transferase (OGT), a eukaryotic glycosyltransferase that utilizes UDP-GlcNAc as a glycosyl donor.

  • Biological active peptide
  • Substrate of O-linked GlcNAc transferase (OGT)
  • White to off-white solid appearance
  • High purity of 98.25%
  • Soluble in DMSO and H2O at 100 mg/mL
  • Sequence: Lys-Lys-Lys-Tyr-Pro-Gly-Gly-Ser-Thr-Pro-Val-Ser-Ser-Ala-Asn-Met-Met

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.