Learn More
Medchemexpress LLC O-linked GlcNAc transferase substrate | 954381-93-2 | 98.3% | 1783.08 | 1 MG
SDP
Catalog No. 5000397992
Encompass_Preferred
O-Linked GlcNAc transferase substrate is a biologically active peptide that functions as a substrate for O-linked GlcNAc transferase (OGT). OGT is a eukaryotic glycosyltransferase that utilizes UDP-GlcNAc as a glycosyl donor. This product is intended for research use only.
  • Biologically active peptide
  • Functions as a substrate for OGT
  • Utilizes UDP-GlcNAc as a glycosyl donor
  • Suitable for research applications
Catalog No. 50-003-97992 Supplier Medchemexpress LLC Supplier No. HYP53751MG
O-Linked GlcNAc transferase substrate is a biologically active peptide that functions as a substrate for O-linked GlcNAc transferase (OGT). OGT is a eukaryotic glycosyltransferase that utilizes UDP-GlcNAc as a glycosyl donor. This product is intended for research use only.
  • Biologically active peptide
  • Functions as a substrate for OGT
  • Utilizes UDP-GlcNAc as a glycosyl donor
  • Suitable for research applications