Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Methyl-L-tyrosine 98.0+%, TCI America™
SDP

Catalog No. M22761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22761G Supplier TCI America Supplier No. M22761G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent used for research applications.

Chemical Identifiers

CAS 6230-11-1
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002604
InChI Key GEYBMYRBIABFTA-IMWMWJONNA-N
Synonym 4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh
PubChem CID 2723935
IUPAC Name (2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C[C@H](N)C(O)=O)C=C1

Specifications

Melting Point 245°C
Color White-Yellow
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥98.0% (HPLC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material O-Methyl-L-tyrosine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.