Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific O-Methylhydroxylamine hydrochloride, 593-56-6, MFCD00012951, 25g

Catalog No. 50424293
Encompass_Preferred

Molecular Formula CH6ClNO, Purity 95+%, Molecular Weight 83.52, Melting Point ca 148 dec.°

Catalog No. 50-424-293 Supplier Matrix Scientific Supplier No. 07503425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula CH6ClNO, Purity 95+%, Molecular Weight 83.52, Melting Point ca 148 dec.°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.