Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI O-Methylhydroxylamine Hydrochloride, >97.0% Purity, Powder, 500 g

Catalog No. 502781483
Encompass_Preferred

O-Methylhydroxylamine hydrochloride, also known as methoxyamine hydrochloride, is a chemical reagent supplied as a white to almost white powder or crystal. This compound is used in various organic synthesis applications.

  • Purity: ≥97.0% (titration)
  • Molecular weight: 83.52
  • Melting point: 149 °C (dec.)
  • Solubility: Soluble in water; slightly soluble in ethanol
  • Storage: Store at room temperature under inert gas
  • Safety: Hygroscopic; classified as corrosive

Catalog No. 50-278-1483 Supplier Grainger Supplier No. 19KW69
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-Methylhydroxylamine hydrochloride, also known as methoxyamine hydrochloride, is a chemical reagent supplied as a white to almost white powder or crystal. This compound is used in various organic synthesis applications.

  • Purity: ≥97.0% (titration)
  • Molecular weight: 83.52
  • Melting point: 149 °C (dec.)
  • Solubility: Soluble in water; slightly soluble in ethanol
  • Storage: Store at room temperature under inert gas
  • Safety: Hygroscopic; classified as corrosive

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.