Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich o-Methylisourea bisulfate

Catalog No. M5370125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M5370125G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M5370125G Supplier Sigma Aldrich Supplier No. M5370125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

o-Methylisourea bisulfate

Specifications

CAS 29427-58-5
Melting Point 118°C to 120°C (lit.)
Molecular Formula C2H6N2O -+ H2SO4
Linear Formula H2NC(OCH3)=NH -+ H2SO4
MDL Number MFCD00013132
Quantity 1 g
Synonym 2-Methylpseudourea monosulfate (1:1); OMI™
Molecular Weight (g/mol) 172.16
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.