Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O=[N+](C1=CC2=C(N(CC3=CC=CC=C3)N=C2OCC4CCCCC4)C=C1)[O-] | 1564253-75-3 | 99.7% | 365.43 | C21H23N3O3 | 5 MG
SDP

Catalog No. 5000222128
Encompass_Preferred

PGN36 is a selective cannabinoid CB2 receptor antagonist used as a research tool in neurobiology and bone biology. It shows high affinity for CB2R and is reported to cross the blood-brain barrier, making it suitable for in vivo and in vitro studies of neurodegeneration and collagen type I regulation.

  • Selective CB2 receptor antagonist (Ki 0.09 μM)
  • Marked selectivity over CB1 receptor (>40 μM)
  • Used in studies of frontotemporal dementia and bone-related assays
  • Molecular formula C21H23N3O3; molecular weight 365.43 g·mol-1
  • Purity 99.74%
  • Appearance light yellow solid
  • Storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month
  • Available in small research pack sizes such as 5 mg

Catalog No. 50-002-22128 Supplier Medchemexpress LLC Supplier No. HY1461345MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PGN36 is a selective cannabinoid CB2 receptor antagonist used as a research tool in neurobiology and bone biology. It shows high affinity for CB2R and is reported to cross the blood-brain barrier, making it suitable for in vivo and in vitro studies of neurodegeneration and collagen type I regulation.

  • Selective CB2 receptor antagonist (Ki 0.09 μM)
  • Marked selectivity over CB1 receptor (>40 μM)
  • Used in studies of frontotemporal dementia and bone-related assays
  • Molecular formula C21H23N3O3; molecular weight 365.43 g·mol-1
  • Purity 99.74%
  • Appearance light yellow solid
  • Storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month
  • Available in small research pack sizes such as 5 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.